Learn More
Bismuth(III) selenide, 99.995% (metals basis), Thermo Scientific™
49.70€ - 192.00€
Chemical Identifiers
| CAS | 12068-69-8 |
|---|---|
| MDL Number | MFCD00014200 |
| Synonym | dibismuth selenium 2-, dibismuth triselenide, dibismuth 3+ ion triselenide, bismuth selenide,, bismuth iii selenide trace metals basis 10g |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15482907
|
Thermo Scientific Alfa Aesar
047198.03 |
1g |
49.70€
1g |
Estimated Shipment: 05-10-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15492907
|
Thermo Scientific Alfa Aesar
047198.06 |
5g |
192.00€
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 12068-69-8 | |
| MFCD00014200 | |
| dibismuth selenium 2-, dibismuth triselenide, dibismuth 3+ ion triselenide, bismuth selenide,, bismuth iii selenide trace metals basis 10g |
Specifications
| 12068-69-8 | |
| UN3283 | |
| dibismuth selenium 2-, dibismuth triselenide, dibismuth 3+ ion triselenide, bismuth selenide,, bismuth iii selenide trace metals basis 10g | |
| 654.84 | |
| 6.82 g/mL | |
| Bismuth(III) selenide |
| MFCD00014200 | |
| 14,1278 | |
| Insoluble in water. | |
| 99.999% | |
| (metals basis) |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
missing translation for 'einecsNumber' : 235-104-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Bismuth(III) selenide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P403-Store in a well-ventilated place.
- P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.