accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 120923-37-7

CH3OOCH3NNHONHSOONCH3SH3COON

CAS RN 120923-37-7

IUPAC Name: 1-(4,6-dimethoxypyrimidin-2-yl)-3-[methanesulfonyl(methyl)sulfamoyl]urea
Synonyms: amidosulfuron
Molecular Weight (g/mol): 369.37
Molecular Formula: C9H15N5O7S2
InChi Key: CTTHWASMBLQOFR-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(NC(=O)NS(=O)(=O)N(C)S(C)(=O)=O)=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results