CAS RN 120964-45-6
IUPAC Name:
5-{4-amino-1H-imidazo[4,5-c]pyridin-1-yl}-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol hydrochloride
Synonyms:
5-{4-aminoimidazo[4,5-c]pyridin-1-yl}-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol hydrochloride
Molecular Weight (g/mol):
298.73
Molecular Formula:
C12H15ClN4O3
InChi Key:
UNSKMHKAFPRFTI-UHFFFAOYNA-N
SMILES:
Cl.NC1=C2N=CN(C3C=C(CO)C(O)C3O)C2=CC=N1
1
–
3
of
3
results
1
Promotions Available
Tocris Bioscience™ DZNep HCl
Tocris Bioscience™ DZNep HCl
3-Deazaneplanocin A hydrochloride synthesized to Ancillary Material Grade
2
3-Deazaneplanocin A Hydrochloride, TRC
CAS: 120964-45-6 Molecular Formula: C12 H14 N4 O3 . Cl H Molecular Weight (g/mol): 298.73 IUPAC Name: (1S,2R,5R)-5-(4-aminoimidazo[4,5-c]pyridin-1-yl)-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol;hydrochloride SMILES: Cl.Nc1nccc2c1ncn2[C@@H]3C=C(CO)[C@@H](O)[C@H]3O
3
Promotions Available
Tocris Bioscience™ 3-Deazaneplanocin A hydrochloride
Tocris Bioscience™ 3-Deazaneplanocin A hydrochloride