accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOClFFF

CAS RN 121-17-5

IUPAC Name: 1-chloro-2-nitro-4-(trifluoromethyl)benzene
Synonyms: 1-chloro-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: TZGFQIXRVUHDLE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Concentration 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)