missing translation for 'onlineSavingsMsg'
Learn More

5-Androstane in DCM 1000μg/mL, Fisher Chemical™

Product Code. 12917093 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12917093 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12917093 Supplier Fisher Chemical Supplier No. P882210

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Bottle
CAS 121-17-5
Analyte or Component Names 5_-Androstane
Molecular Formula C7H3ClF3NO2
Concentration 1000 μg/mL
MDL Number MFCD00007084
InChI Key TZGFQIXRVUHDLE-UHFFFAOYSA-N
Type Mass Spectrometry Standard
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
Element 5-Androstane
IUPAC Name 1-chloro-2-nitro-4-(trifluoromethyl)benzene
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 225.55
PubChem CID 8462
Quantity 5 mL
Chemical Name or Material Aminohippuric acid reagent.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.