accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121-39-1

IUPAC Name: ethyl 3-phenyloxirane-2-carboxylate
Synonyms: ethyl 3-phenylglycidate
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: GOMAKLPNAAZVCJ-UHFFFAOYNA-N
SMILES: CCOC(=O)C1OC1C1=CC=CC=C1
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)