accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 121-54-0

IUPAC Name: benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
Synonyms: benzethonium chloride 苄索氯铵
Molecular Weight (g/mol): 448.09
Molecular Formula: C27H42ClNO2
InChi Key: UREZNYTWGJKWBI-UHFFFAOYSA-M
SMILES: [Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (7)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)