missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride, TRC

Product Code. 30132986
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132986 50 g 50g
30041182 1 g 1g
30041024 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132986 Supplier Toronto Research Chemicals Supplier No. TRCB19025050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27 H42 N O2 . Cl
Molecular Weight (g/mol) 448.08
Synonym Benzyldimethyl[2-[2-[p-(1,1,3,3-tetramethylbutyl)phenoxy]ethoxy]ethyl]ammonium chloride, N,N-Dimethyl-N-[2-[2-[4-(1,1,3,3-tetramethylbutyl)phenoxy]ethoxy]ethyl]benzenemethanaminium chloride, (Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, 2-[2-[p-(Diisobutyl)phenoxy]ethoxy]ethyldimethylbenzylammonium chloride, Anti-Germ 77, Antiseptol, Antiseptol (quaternary compound), BZT, Bencetonium chloride, Benzethionium chloride, Benzethonium chloride, Benzetonium chloride, Benzyldimethyl[2-[2-[4-(1,1,3,3-tetramethylbutyl)phenoxy]ethoxy]ethyl]ammonium chloride, Benzyldimethyl[2-[2-[p-(1,1,3,3-tetramethylbutyl)phenoxy]ethoxy]ethyl]ammonium chloride, Diapp, Hyamine 1622, LonzaGuard, MED 81, N-Benzyl-N,N-dimethyl-N-(4-[1,1,3,3-tetramethylbutyl]phenoxyethoxyethyl)ammonium chloride, Phemeride, Phemerol, Phemerol chloride, Phemithyn, Polymine D, Quatrachlor, Solamin, [2-[2-(4-Diisobutylphenoxy)ethoxy]ethyl]dimethylbenzylammonium chloride, p-tert-Octylphenoxyethoxyethyldimethylbenzylammonium chloride
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES [Cl-].CC(C)(C)CC(C)(C)c1ccc(OCCOCC[N+](C)(C)Cc2ccccc2)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C27H42NO2.ClH/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H/q+1;/p-1
Formula Weight 447.2904
Chemical Name or Material Benzethonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.