CAS RN 1212-53-9
CAS RN 1212-53-9
IUPAC Name:
methyl 2-{[(benzyloxy)carbonyl]amino}acetate
Synonyms:
methyl 2-{[(benzyloxy)carbonyl]amino}acetate
Molecular Weight (g/mol):
223.23
Molecular Formula:
C11H13NO4
InChi Key:
DZYBBBYFLOPVOL-UHFFFAOYSA-N
SMILES:
COC(=O)CNC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N-Benzyloxycarbonylglycine methyl ester, 98%
CAS: 1212-53-9 Molecular Formula: C11H13NO4 Molecular Weight (g/mol): 223.23 MDL Number: MFCD00043631 InChI Key: DZYBBBYFLOPVOL-UHFFFAOYSA-N Synonym: z-gly-ome,z-glycine methyl ester,methyl 2-benzyloxy carbonyl amino acetate,carbobenzyloxyglycine methyl ester,n-cbz-glycine methyl ester,methyl n-benzyloxycarbonylglycinate,cbz-glycine methyl ester,carbobenzoxyglycine methyl ester,methyl 2-benzyloxycarbonylamino acetate,glycine, n-phenylmethoxy carbonyl-, methyl ester PubChem CID: 71033 SMILES: COC(=O)CNC(=O)OCC1=CC=CC=C1