missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonylglycine methyl ester, 98%

Product Code. 15489308
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489308 25 g 25g
15479308 5 g 5g
15499308 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15489308 Supplier Thermo Scientific Alfa Aesar Supplier No. H62858.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1212-53-9 | C11H13NO4 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1212-53-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00043631
InChI Key DZYBBBYFLOPVOL-UHFFFAOYSA-N
Synonym z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester
PubChem CID 71033
SMILES COC(=O)CNC(=O)OCC1=CC=CC=C1

Spécification

Melting Point 22°C to 26°C
Boiling Point 147°C to 151°C
Linear Formula C6H5CH2OCONHCH2CO2CH3
Quantity 25 g
IUPAC Name methyl 2-{[(benzyloxy)carbonyl]amino}acetate
Formula Weight 223.23
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonylglycine methyl ester

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.