accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH4Br

CAS RN 12124-97-9

IUPAC Name: ammonium bromide
Synonyms: ammonium bromide 阿扎溴铵
Molecular Weight (g/mol): 97.94
Molecular Formula: BrH4N
InChi Key: SWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES: [NH4+].[Br-]
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
Grade 
  • (4)
  • (4)
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (4)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (9)
  • (10)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (4)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (4)