missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bromide, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10161750 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10161750 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10161750 Supplier Fisher Chemical Supplier No. A/3520/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 235°C
Melting Point 452°C
CAS 12124-97-9
Color White
pH 4.5 to 5.5
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula BrH4N
MDL Number 11419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES [NH4+].[Br-]
IUPAC Name ammonium bromide
Molecular Weight (g/mol) 97.94
Formula Weight 97.94g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.