accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHN

CAS RN 1215-59-4

IUPAC Name: 5-(benzyloxy)-1H-indole
Synonyms: indole, 5-benzyloxy-
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: JCQLPDZCNSVBMS-UHFFFAOYSA-N
SMILES: C(OC1=CC=C2NC=CC2=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)