accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHOHN

CAS RN 122321-04-4

IUPAC Name: 2-[(2-hydroxyethyl)(methyl)amino]pyridin-1-ium
Synonyms: 2-[(2-hydroxyethyl)(methyl)amino]pyridin-1-ium
Molecular Weight (g/mol): 153.2
Molecular Formula: C8H13N2O
InChi Key: MWGKOPUDDQZERY-UHFFFAOYSA-O
SMILES: CN(CCO)C1=CC=CC=[NH+]1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results