accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFN

CAS RN 122376-76-5

IUPAC Name: 2-(3,5-difluorophenyl)acetonitrile
Synonyms: 2-(3,5-difluorophenyl)acetonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: OBMYMTSBABEPIB-UHFFFAOYSA-N
SMILES: FC1=CC(CC#N)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results