accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONClNBrFFF

CAS RN 122453-73-0

IUPAC Name: 4-bromo-2-(4-chlorophenyl)-1-(ethoxymethyl)-5-(trifluoromethyl)-1H-pyrrole-3-carbonitrile
Synonyms: pylon chlorfenpyr pirate 3F chlorfenapyr C15H11BrClF3N2O pirate kotetsu
Molecular Weight (g/mol): 407.62
Molecular Formula: C15H11BrClF3N2O
InChi Key: CWFOCCVIPCEQCK-UHFFFAOYSA-N
SMILES: CCOCN1C(=C(C#N)C(Br)=C1C(F)(F)F)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results