missing translation for 'onlineSavingsMsg'
Learn More

Chlorfenapyr, TRC

Product Code. 30137238
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137238 250 mg 250mg
30133607 1 g 1g
30040867 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137238 Supplier Toronto Research Chemicals Supplier No. TRCC428500250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122453-73-0
Molecular Formula C15 H11 Br Cl F3 N2 O
Molecular Weight (g/mol) 407.61
Synonym 4-Bromo-2-(4-chlorophenyl)-1-(ethoxymethyl)-5-(trifluoromethyl)-1H-pyrrole-3-carbonitrile, 4-Bromo-2-(p-chlorophenyl)-1-(ethoxymethyl)-5-(trifluoromethyl)pyrrole-3-carbonitrile, AC 303-630, AC 303630, Bromchlorfenapyr, CL 303630, Chlorfenapyr, Citrex, Citrex (acaricide), Kotetsu, Lepido, Mystox MP, Phantom, Pirate, Pirate 3F, Pylon, Rampage, Rampage (insecticide), Secure, Secure 360SC Insecticide-Miticide, Stealth
IUPAC Name 4-bromo-2-(4-chlorophenyl)-1-(ethoxymethyl)-5-(trifluoromethyl)pyrrole-3-carbonitrile
SMILES CCOCn1c(c2ccc(Cl)cc2)c(C#N)c(Br)c1C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H11BrClF3N2O/c1-2-23-8-22-13(9-3-5-10(17)6-4-9)11(7-21)12(16)14(22)15(18,19)20/h3-6H,2,8H2,1H3
Formula Weight 405.9695
Percent Purity >95%
Chemical Name or Material Chlorfenapyr
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.