accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3O

CAS RN 123-05-7

IUPAC Name: 2-ethylhexanal
Synonyms: 2-ethylhexanal ethylhexanal
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: LGYNIFWIKSEESD-UHFFFAOYNA-N
SMILES: CCCCC(CC)C=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)