accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO

CAS RN 123-08-0

IUPAC Name: 4-hydroxybenzaldehyde
Synonyms: P-hydroxybenzaldehyde parahydroxybenzaldehyde usaf M-6 hydroxybenzaldehyde
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: RGHHSNMVTDWUBI-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (9)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (9)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)