accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 123-38-6

IUPAC Name: propanal
Synonyms: propionaldehyde proprionaldehyde C2H5cho fema number 2923 propionaldehyde, 1-14c-labeled 1-14C-labeled propionaldehyde propionaldehyde (substance) 2-14C-labeled propionaldehyde propionaldehyde, 2-14C-labeled propionaldehyde (natural)
Molecular Weight (g/mol): 58.08
Molecular Formula: C3H6O
InChi Key: NBBJYMSMWIIQGU-UHFFFAOYSA-N
SMILES: CCC=O
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)
  • (1)
  • (9)
Percent Purity 
  • (1)
  • (2)
  • (7)
  • (4)
Physical Form 
  • (4)
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (9)
  • (3)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (9)
  • (1)
  • (2)
  • (7)
  • (4)
  • (4)
  • (1)
  • (5)
  • (1)