accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH3CNOOClOHNN

CAS RN 123040-16-4

IUPAC Name: hydrogen N-{1-azabicyclo[2.2.2]octan-3-yl}-6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxamide chloride
Synonyms: hydrogen azasetron chloride
Molecular Weight (g/mol): 386.27
Molecular Formula: C17H21Cl2N3O3
InChi Key: DBMKBKPJYAHLQP-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN1C(=O)COC2=C1C=C(Cl)C=C2C(=O)NC1CN2CCC1CC2
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results