missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Azasetron hydrochloride

Product Code. 16677022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16677022 250 mg 250mg
16667022 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16677022 Supplier Thermo Scientific Chemicals Supplier No. 465072500
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123040-16-4
Molecular Formula C17 H20 Cl N3 O3 . H Cl

Specifications

Quantity 250 mg
Formula Weight 386.27
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key DBMKBKPJYAHLQP-UHFFFAOYNA-N
SMILES [H+].[Cl-].CN1C(=O)COC2=C1C=C(Cl)C=C2C(=O)NC1CN2CCC1CC2
IUPAC Name hydrogen N-{1-azabicyclo[2.2.2]octan-3-yl}-6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxamide chloride
Molecular Weight (g/mol) 386.27
Chemical Name or Material Azasetron hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.