accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONNNOHNH2(E/Z)

CAS RN 1233243-62-3

IUPAC Name: tert-butyl 4-[4-(N'-hydroxycarbamimidoyl)phenyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[4-(N'-hydroxycarbamimidoyl)phenyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 320.39
Molecular Formula: C16H24N4O3
InChi Key: XNXLELZXYPNZRJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)C(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results