accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 123639-61-2

OOOHONHOO

CAS RN 123639-61-2

IUPAC Name: 5-(benzyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
Synonyms: 5-(benzyloxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxopentanoic acid
Molecular Weight (g/mol): 459.5
Molecular Formula: C27H25NO6
InChi Key: HJJURMMMGPQIQP-UHFFFAOYNA-N
SMILES: OC(=O)C(CCC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results