Learn More
N-Fmoc-L-glutamic acid 5-benzyl ester, 95%, Thermo Scientific™
Product Code. 15499258
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15499258
Brand: Thermo Scientific Alfa Aesar H62783.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123639-61-2
Molecular Formula C27H25NO6
Molecular Weight (g/mol) 459.498
MDL Number MFCD00065642
InChI Key HJJURMMMGPQIQP-DEOSSOPVSA-N
Synonym fmoc-glu obzl-oh, fmoc-l-glutamic acid-gamma-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid 5-benzyl ester, 2s-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, pubchem10013, fmoc-glu obzl-oh hplc, n-9-fluorenylmethoxycarbonyl-l-glutamic acid-gamma-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-benzyl ester, 2s-2-fluoren-9-ylmethoxy carbonylamino-4-benzyloxycarbonyl butanoic acid
PubChem CID 13966932
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Specifications
Melting Point (decomposition)
Quantity 25 g
Beilstein 3576722
Optical Rotation +12° (c=1 in Chloroform)
Formula Weight 459.49
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-glutamic acid 5-benzyl ester