accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO

CAS RN 124-13-0

IUPAC Name: octanal
Synonyms: octanal oktylaldehyd 1-caprylaldehyde C-8 aldehyde caprylic aldehyde octylaldehyde caprylaldehyd tech. octanal octyl aldehyde (substance) kaprylaldehyd
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: NUJGJRNETVAIRJ-UHFFFAOYSA-N
SMILES: CCCCCCCC=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)