accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH2

CAS RN 124-22-1

IUPAC Name: dodecan-1-amine
Synonyms: N-dodecylamine
Molecular Weight (g/mol): 185.36
Molecular Formula: C12H27N
InChi Key: JRBPAEWTRLWTQC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)