missing translation for 'onlineSavingsMsg'
Learn More

Dodecylamine, TRC

Product Code. 30385849
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385849 Supplier Toronto Research Chemicals Supplier No. TRCD49470525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Dodecylamine
CAS 124-22-1
Molecular Formula C12 H27 N
Quantity 25 g
Synonym 1-Dodecanamine (9CI, ACI), Dodecylamine (8CI), 1-Aminododecane, 1-Dodecylamine, Adogen 163D, Alamine 4, Amine BB, Armeen 12D, Farmin 20D, HAN 12, Kemamine P 690, Lauramine, Laurinamine, Laurylamine, Lipomin 12D, Monododecylamine, Monolaurylamine, n-Dodecylamine, n-Laurylamine, Nissan Amine BB
InChI Formula InChI=1S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3
SMILES CCCCCCCCCCCCN
IUPAC Name dodecan-1-amine
Molecular Weight (g/mol) 185.35
Formula Weight 185.2143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.