accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSFFFFFF

CAS RN 1240257-57-1

IUPAC Name: 1-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]ethan-1-one
Synonyms: 1-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]ethanone
Molecular Weight (g/mol): 264.19
Molecular Formula: C8H6F6OS
InChi Key: XABWPYOWUIIPAB-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results