accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHOHN

CAS RN 124035-23-0

IUPAC Name: N-[4-(piperidine-4-carbonyl)phenyl]acetamide
Synonyms: N-[4-(piperidine-4-carbonyl)phenyl]acetamide
Molecular Weight (g/mol): 246.31
Molecular Formula: C14H18N2O2
InChi Key: LLEYCMVEUNZRIO-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C(=O)C1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results