accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOH3COOHCH3HOCH3O

CAS RN 125-46-2

IUPAC Name: 4,10-diacetyl-11,13-dihydroxy-2,12-dimethyl-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(13),6,9,11-tetraene-3,5-dione
Synonyms: 7-hydroxy-(S)-usnate
Molecular Weight (g/mol): 344.32
Molecular Formula: C18H16O7
InChi Key: CUCUKLJLRRAKFN-UHFFFAOYNA-N
SMILES: CC(=O)C1C(=O)C=C2OC3=C(C(C)=O)C(O)=C(C)C(O)=C3C2(C)C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results