CAS RN 1254944-66-5
CAS RN 1254944-66-5
IUPAC Name:
ethyl 2-[5-(methylsulfanyl)thiophene-2-carbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms:
ethyl 2-[5-(methylsulfanyl)thiophene-2-carbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylate
Molecular Weight (g/mol):
361.47
Molecular Formula:
C18H19NO3S2
InChi Key:
UIEBLUZPSFAFOC-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1CC2=CC=CC=C2CN1C(=O)C1=CC=C(SC)S1
1
–
2
of
2
results
1
Tocris Bioscience™ SR 8278
CAS: 1254944-66-5 Molecular Formula: C18H19NO3S2
2
SR 8278, TRC
CAS: 1254944-66-5 Molecular Formula: C18H19NO3S2 Molecular Weight (g/mol): 361.48 Synonym: SR 8278,1,2,3,4-Tetrahydro-2-[[5-(methylthio)-2-thienyl]carbonyl]-3-isoquinolinecarboxylic Acid Ethyl Ester IUPAC Name: ethyl 2-(5-methylsulfanylthiophene-2-carbonyl)-3,4-dihydro-1H-isoquinoline-3-carboxylate SMILES: CCOC(=O)C1Cc2ccccc2CN1C(=O)c3ccc(SC)s3