missing translation for 'onlineSavingsMsg'
Learn More

SR 8278, TRC

Product Code. 30428286
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428286 50 mg
30418246 100 mg
30426810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428286 Supplier Toronto Research Chemicals Supplier No. TRCS68425050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1254944-66-5
Molecular Formula C18H19NO3S2
Molecular Weight (g/mol) 361.48
Synonym SR 8278, 1,2,3,4-Tetrahydro-2-[[5-(methylthio)-2-thienyl]carbonyl]-3-isoquinolinecarboxylic Acid Ethyl Ester
IUPAC Name ethyl 2-(5-methylsulfanylthiophene-2-carbonyl)-3,4-dihydro-1H-isoquinoline-3-carboxylate
SMILES CCOC(=O)C1Cc2ccccc2CN1C(=O)c3ccc(SC)s3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H19NO3S2/c1-3-22-18(21)14-10-12-6-4-5-7-13(12)11-19(14)17(20)15-8-9-16(23-2)24-15/h4-9,14H,3,10-11H2,1-2H3
Formula Weight 361.0806
Chemical Name or Material 1,2,3,4-Tetrahydro-2-((5-(methylthio)-2-thienyl)carbonyl)-3-isoquinolinecarboxylic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.