accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NHNNOHCl

CAS RN 1257326-24-1

IUPAC Name: 1-phenyl-3-[(piperidin-2-yl)methyl]imidazolidin-2-one hydrochloride
Synonyms: 1-phenyl-3-(piperidin-2-ylmethyl)imidazolidin-2-one hydrochloride
Molecular Weight (g/mol): 295.81
Molecular Formula: C15H22ClN3O
InChi Key: BYJUVPWUMBVSTK-UHFFFAOYNA-N
SMILES: Cl.O=C1N(CC2CCCCN2)CCN1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results