missing translation for 'onlineSavingsMsg'
Learn More

GSK 789472 Hydrochloride, TRC

Product Code. 30240996
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240996 10 mg 10mg
30240818 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240996 Supplier Toronto Research Chemicals Supplier No. TRCG79780510MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257326-24-1
Molecular Formula C15H21N3O . HCl
Molecular Weight (g/mol) 295.81
Synonym GSK 789472 Hydrochloride, 1-Phenyl-3-(2-piperidinylmethyl)-2-imidazolidinone Hydrochloride;
IUPAC Name 1-phenyl-3-(piperidin-2-ylmethyl)imidazolidin-2-one;hydrochloride
SMILES Cl.O=C1N(CC2CCCCN2)CCN1c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H21N3O.ClH/c19-15-17(12-13-6-4-5-9-16-13)10-11-18(15)14-7-2-1-3-8-14;/h1-3,7-8,13,16H,4-6,9-12H2;1H
Formula Weight 295.1451
Chemical Name or Material 1-Phenyl-3-(2-piperidinylmethyl)-2-imidazolidinone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.