accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1263283-43-7

H3CNONHONOHNOHCl

CAS RN 1263283-43-7

IUPAC Name: 4-[2-(3,5-dioxopiperazin-1-yl)propyl]piperazine-2,6-dione hydrochloride
Synonyms: razoxane hydrochloride
Molecular Weight (g/mol): 304.73
Molecular Formula: C11H17ClN4O4
InChi Key: BIFMNMPSIYHKDN-UHFFFAOYNA-N
SMILES: Cl.CC(CN1CC(=O)NC(=O)C1)N1CC(=O)NC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results