missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Dexrazoxane hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Topoisomerase II inhibitor and intracellular ion chelator. Bridges and stabilizes an interface between two ATPase promoters to inhibit topoisomerase II activity. Cardioprotective when co-administered with doxorubicin; decreases formation of reactive oxygen species (ROS) and activates the PI3K/Akt survival pathway.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 304.73 |
| Purity | >98% |
| Molecular Weight (g/mol) | 304.731 |
| Chemical Name or Material | Dexrazoxane hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction