accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1266615-56-8

IUPAC Name: sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate dihydrate
Synonyms: sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzenolate dihydrate
Molecular Weight (g/mol): 326.1
Molecular Formula: C12H10Cl2NNaO4
InChi Key: WZHXLAHEFATELD-UHFFFAOYSA-M
SMILES: O.O.[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (4)
  • (3)
Color 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results