accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 127-17-3

IUPAC Name: 2-oxopropanoic acid
Synonyms: pyruvic acid acid, pyruvic 2-oxopropionsaeure BTS pyruvic acid (substance) pyroracemate brenztraubensaeure ion(1-) 2-oxopropanoic acid α-oxopropionsaeure 2-oxopropanoate
Molecular Weight (g/mol): 88.06
Molecular Formula: C3H4O3
InChi Key: LCTONWCANYUPML-UHFFFAOYSA-N
SMILES: CC(=O)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results