CAS RN 1273-86-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ferrocenemethanol, 97%, Thermo Scientific Chemicals
CAS: 1273-86-5 Molecular Formula: C11H12FeO Molecular Weight (g/mol): 216.06 MDL Number: MFCD00040878 InChI Key: KLMBXXGIFRROFL-UHFFFAOYSA-N SMILES: [Fe].c1cccc1.OCc1cccc1