missing translation for 'onlineSavingsMsg'
Learn More

Ferrocenemethanol, 97%, Thermo Scientific Chemicals

Product Code. 10554921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10554921 1 g Glass bottle 1g
10510784 500 mg Glass bottle 500mg
10410062 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10554921 Supplier Thermo Scientific Acros Supplier No. 348030010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1273-86-5
Molecular Formula C11H12FeO
Molecular Weight (g/mol) 216.06
MDL Number MFCD00040878
InChI Key KLMBXXGIFRROFL-UHFFFAOYSA-N
SMILES [Fe].c1cccc1.OCc1cccc1

Specifications

Melting Point 77.0°C to 82.0°C
Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 215.93
Percent Purity 97%
Physical Form Needles or Powder
Chemical Name or Material Ferrocenemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.