accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CCCY

CAS RN 1294-07-1

IUPAC Name: yttrium(3+) tris(cyclopenta-1,3-dien-1-ide)
Synonyms: yttrium(3+) tris(cyclopenta-1,3-dien-1-ide)
Molecular Weight (g/mol): 284.19
Molecular Formula: C15H15Y
InChi Key: IUJOPJIXAVSZIQ-UHFFFAOYSA-N
SMILES: [Y+3].C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)