accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 129648-07-3

CH3PPCH3CH3H3C

CAS RN 129648-07-3

IUPAC Name: 1-[2-(2,5-dimethylphospholan-1-yl)ethyl]-2,5-dimethylphospholane
Synonyms: 1-[2-(2,5-dimethylphospholan-1-yl)ethyl]-2,5-dimethylphospholane
Molecular Weight (g/mol): 258.33
Molecular Formula: C14H28P2
InChi Key: IRCDUOCGSIGEAI-UHFFFAOYNA-N
SMILES: CC1CCC(C)P1CCP1C(C)CCC1C
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results