missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane, 97+%, Thermo Scientific™

Product Code. 15455456
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455456 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15455456 Supplier Thermo Scientific Alfa Aesar Supplier No. H60071.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129648-07-3
Molecular Formula C14H28P2
Molecular Weight (g/mol) 258.326
MDL Number MFCD01073770
InChI Key IRCDUOCGSIGEAI-AAVRWANBSA-N
Synonym r,r-me-bpe, unii-1ls1afh1f1, 1ls1afh1f1, +-1,2-bis 2r,5r-2,5-dimethylphospholano ethane, s,s-me-en-duphos, me-bpe, r,r, r,r-me-bpe mi, r,r-me-bpe, +, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl ethyl-2,5-dimethylphospholane
PubChem CID 2734555
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]ethyl]-2,5-dimethylphospholane
SMILES CC1CCC(P1CCP2C(CCC2C)C)C

Specifications

Density 0.94
Boiling Point 64°C to 67°C (0.06 mmHg)
Refractive Index 1.534
Quantity 1 g
UN Number UN2845
Sensitivity Air Sensitive
Formula Weight 258.32
Percent Purity ≥97%
Chemical Name or Material 1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.