accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1299-86-1

AlAlC(?)AlAl

CAS RN 1299-86-1

IUPAC Name: 1,3,5,7-tetraaluminahepta-3,4-dien-1,6-diyne
Synonyms: aluminium carbide
Molecular Weight (g/mol): 143.96
Molecular Formula: C3Al4
InChi Key: CAVCGVPGBKGDTG-UHFFFAOYSA-N
SMILES: [Al]#C[Al]=C=[Al]C#[Al]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results