accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOO2H2H2H

CAS RN 13031-32-8

IUPAC Name: nitro(²H₃)methane
Synonyms: nitro(²H₃)methane
Molecular Weight (g/mol): 64.06
Molecular Formula: CH3NO2
InChi Key: LYGJENNIWJXYER-FIBGUPNXSA-N
SMILES: [2H]C([2H])([2H])[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)