accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHCa

CAS RN 1305-62-0

IUPAC Name: calcium dihydroxide
Synonyms: calcium hydroxide hydralime Ca(OH)2 calcium hydrate hydrated lime loeschkalk kalziumhydroxid slaked lime geloeschter kalk
Molecular Weight (g/mol): 74.09
Molecular Formula: CaH2O2
InChi Key: AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Ca++]
Quantity 
  • (7)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
Grade 
  • (2)
  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
Physical Form 
  • (7)
  • (1)
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (4)
  • (5)
  • (1)
  • (9)
  • (5)
  • (3)
  • (2)
  • (7)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (7)
  • (1)
  • (3)
  • (5)