accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13080-86-9

CH3H3COH2NONH2

CAS RN 13080-86-9

IUPAC Name: 4-(4-{2-[4-(4-aminophenoxy)phenyl]propan-2-yl}phenoxy)aniline
Synonyms: 4-(4-{2-[4-(4-aminophenoxy)phenyl]propan-2-yl}phenoxy)aniline
Molecular Weight (g/mol): 410.52
Molecular Formula: C27H26N2O2
InChi Key: KMKWGXGSGPYISJ-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=C(OC2=CC=C(N)C=C2)C=C1)C1=CC=C(OC2=CC=C(N)C=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results