accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOFFF

CAS RN 13081-18-0

IUPAC Name: ethyl 3,3,3-trifluoro-2-oxopropanoate
Synonyms: ethyl 3,3,3-trifluoro-2-oxopropanoate
Molecular Weight (g/mol): 170.09
Molecular Formula: C5H5F3O3
InChi Key: KJHQVUNUOIEYSV-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)