accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3

CAS RN 131-18-0

IUPAC Name: 1,2-dipentyl benzene-1,2-dicarboxylate
Synonyms: amoil DPNP DPP di-n-pentylphthalate di-N-amyl phthalate
Molecular Weight (g/mol): 306.4
Molecular Formula: C18H26O4
InChi Key: IPKKHRVROFYTEK-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results